Gene Transcripts

Gene Symbol
Trpm5
Entrez Gene ID
56843
Gene Name
transient receptor potential cation channel, subfamily M, member 5
Gene Aliases
9430099A16Rik, LTrpC-5, Ltrpc5, Mtr1
Chromosome Location
Chr. 7: 143071529 - 143094642 on Build GRCm38
UniGene ID
Mm.286668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020277.2 NP_064673.2 22 3253
GenBank mRNA AB039952.1 - 21 3170
AF228681.1 - 22 3253
AJ271092.2 - 21 3166
AK029187.1 - 2 483
AK035197.1 - 18 2783
AK044908.1 - 18 2868
AK161030.1 - 13 1846
AY280364.1 - 22 3217
AY280365.1 - 23 3303
BC133712.1 - 21 3170 40143371

View Full Product Details