Gene Transcripts

Gene Symbol
Tas2r108
Entrez Gene ID
57253
Gene Name
taste receptor, type 2, member 108
Gene Aliases
T2R108, T2r8, Tas2r4, Tas2r8, mGR08, mt2r8
Chromosome Location
Chr. 6: 40493592 - 40494485 on Build GRCm38
UniGene ID
Mm.377132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020502.1 NP_065248.1 1 713
GenBank mRNA AY916507.1 - 1 713
BC116952.1 - 1 776 40126271

View Full Product Details