Gene Transcripts

Gene Symbol
Ttyh1
Entrez Gene ID
57776
Gene Name
tweety homolog 1 (Drosophila)
Gene Aliases
4930459B04Rik, 6330408P11Rik, TTY1, mTTY1, tty
Chromosome Location
Chr. 7: 4119527 - 4136241 on Build GRCm38
UniGene ID
Mm.29729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB162929.1 - 7, 8 895
AF190699.1 - 7, 8 967
AF190991.1 - 7, 8 967
AK015486.1 - 6, 7 618
AK018148.1 - 7, 8 959
AK031550.1 - 7, 8 995
AK042758.1 - 7, 8 748
AK043998.1 - 7, 8 1015
AK048413.1 - 7, 8 966
AK049903.1 - 6, 7 701
AK081981.1 - 7, 8 969
AK083126.1 - 6, 7 1092
BC046310.1 - 7, 8 953 6307420
BC065694.1 - 7, 8 959 6813365

View Full Product Details