Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077698.1 | NP_001071166.1 | 14 | 1598 | |
NM_019679.2 | NP_062653.2 | 14 | 1598 | ||
GenBank mRNA | AF006466.1 | - | 14 | 1345 | |
AF215666.1 | - | 14 | 1423 | ||
AK131173.1 | - | 8 | 734 | ||
AK158820.1 | - | 2 | 564 |