Gene Transcripts
- Gene Symbol
- Pdzk1
- Entrez Gene ID
- 59020
- Gene Name
- PDZ domain containing 1
- Gene Aliases
- 1700023D20Rik, 2610507N21Rik, 4921513F16Rik, AI267131, AI314638, AL022680, D3Ertd537e, Pdzd1, mPDZK1
- Chromosome Location
- Chr. 3: 96830065 - 96870926 on Build GRCm38
- UniGene ID
- Mm.489677
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146001.1 |
NP_001139473.1
|
5, 6 |
849 |
|
NM_021517.2 |
NP_067492.2
|
5, 6 |
895 |
|
GenBank mRNA |
AF220100.1 |
- |
5, 6 |
834 |
|
AF474247.1 |
- |
4, 5 |
833 |
|
AK006269.1 |
- |
5, 6 |
1369 |
|
AK012070.1 |
- |
5, 6 |
885 |
|
AK014879.1 |
- |
5, 6 |
1152 |
|
AK029752.1 |
- |
5, 6 |
1353 |
|
AK029764.1 |
- |
5, 6 |
1363 |
|
AK165415.1 |
- |
5, 6 |
830 |
|
BC013512.1 |
- |
4, 5 |
818 |
4234949 |
View Full Product Details