Gene Transcripts
- Gene Symbol
- Evc
- Entrez Gene ID
- 59056
- Gene Name
- Ellis van Creveld gene syndrome
- Gene Aliases
- Chromosome Location
- Chr. 5: 37288977 - 37337242 on Build GRCm38
- UniGene ID
- Mm.479678
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ250841.1 |
- |
8 |
1054 |
|
View Full Product Details