Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021560.4 | NP_067535.3 | 1 | 1187 | |
GenBank mRNA | AF218865.1 | - | 1 | 809 | |
AK047201.1 | - | 1 | 921 | ||
AK076228.1 | - | 1 | 1111 | ||
BC053007.1 | - | 1 | 1187 | 5686844 |