Gene Transcripts

Gene Symbol
Ncstn
Entrez Gene ID
59287
Gene Name
nicastrin
Gene Aliases
9430068N19Rik, AA727311, Aph2, D1Dau13e, Kiaa0253, Nct, mKIAA0253
Chromosome Location
Chr. 1: 172066013 - 172082749 on Build GRCm38
UniGene ID
Mm.218203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF240469.1 - 14 1600
BC019998.1 - 14 1595 4458780

View Full Product Details