Gene Transcripts

Gene Symbol
Iigp1
Entrez Gene ID
60440
Gene Name
interferon inducible GTPase 1
Gene Aliases
2900074L10Rik, AI046432, AW111922, Ifgga1, Iigp, Irga6
Chromosome Location
Chr. 18: 60373632 - 60392634 on Build GRCm38
UniGene ID
Mm.261140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146275.1 NP_001139747.1 3 934
NM_021792.4 NP_068564.4 2 751
GenBank mRNA AF194871.1 - 1 646
AJ007971.1 - 2 733
AK089702.1 - 2 763
AK153060.1 - 2 753
BC004649.1 - 2 728 3483102
CT010300.1 - 1 629
DV061261.1 - 2 784

View Full Product Details