Gene Transcripts

Gene Symbol
Kcnq4
Entrez Gene ID
60613
Gene Name
potassium voltage-gated channel, subfamily Q, member 4
Gene Aliases
Chromosome Location
Chr. 4: 120696134 - 120747491 on Build GRCm38
UniGene ID
Mm.249977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081142.1 NP_001074611.1 10 1486
GenBank mRNA AF249747.1 - 2 275
AK040190.1 - 10 1419
AK163659.1 - 1172

View Full Product Details