Gene Transcripts

Gene Symbol
Ngb
Entrez Gene ID
64242
Gene Name
neuroglobin
Gene Aliases
Chromosome Location
Chr. 12: 87097531 - 87102539 on Build GRCm38
UniGene ID
Mm.41395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294308.1 NP_001281237.1 4 1120
NM_022414.2 NP_071859.1 4 1108
GenBank mRNA AK140173.1 - 4 1104
AK165775.1 - 4 914
BC024263.1 - 4 1108 5133442

View Full Product Details