Gene Transcripts

Gene Symbol
Slc15a3
Entrez Gene ID
65221
Gene Name
solute carrier family 15, member 3
Gene Aliases
Ci1, cI-1
Chromosome Location
Chr. 19: 10842544 - 10857915 on Build GRCm38
UniGene ID
Mm.27387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023044.2 NP_075531.2 1 643
GenBank mRNA AF121080.1 - 1 651
AK051941.1 - 1 244
AK171698.1 - 296
BC051940.1 - 1 314 5717926

View Full Product Details