Gene Transcripts

Gene Symbol
Cubn
Entrez Gene ID
65969
Gene Name
cubilin (intrinsic factor-cobalamin receptor)
Gene Aliases
AA408369, AL022750, D2Wsu88e
Chromosome Location
Chr. 2: 13276338 - 13491876 on Build GRCm38
UniGene ID
Mm.313915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081084.2 NP_001074553.1 58, 59 9292
GenBank mRNA AF197159.1 - 6, 7 856
BC026593.1 - 7, 8 1092

View Full Product Details