Gene Transcripts

Gene Symbol
Ypel3
Entrez Gene ID
66090
Gene Name
yippee-like 3 (Drosophila)
Gene Aliases
0610043B10Rik, 1190001G19Rik, Suap
Chromosome Location
Chr. 7: 126776975 - 126780514 on Build GRCm38
UniGene ID
Mm.27114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025347.2 NP_079623.1 2 236
NM_026875.2 NP_081151.2 2 336
GenBank mRNA AB098742.1 - 1 41
AF523353.1 - 2 226
AK002925.1 - 2 221
AK003371.1 - 2 237
AK210119.1 - 1 16
BC009171.1 - 2 210

View Full Product Details