Gene Transcripts
- Gene Symbol
- Ghitm
- Entrez Gene ID
- 66092
- Gene Name
- growth hormone inducible transmembrane protein
- Gene Aliases
- 1010001P14Rik, C77840, MICS1, PTD010, Tmbim5
- Chromosome Location
- Chr. 14: 37120444 - 37135327 on Build GRCm38
- UniGene ID
- Mm.182912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199122.1 |
NP_001186051.1
|
2, 3 |
354 |
|
NM_078478.5 |
NP_510963.1
|
2, 3 |
307 |
|
GenBank mRNA |
AF412297.1 |
- |
2, 3 |
251 |
|
AK005636.1 |
- |
2, 3 |
251 |
|
AK093887.1 |
- |
2, 3 |
283 |
|
AK134592.1 |
- |
2, 3 |
237 |
|
AK146433.1 |
- |
2, 3 |
214 |
|
AK146853.1 |
- |
2, 3 |
270 |
|
AK151488.1 |
- |
2, 3 |
284 |
|
AK158904.1 |
- |
2, 3 |
297 |
|
AK159695.1 |
- |
2, 3 |
281 |
|
AK159724.1 |
- |
2, 3 |
281 |
|
AK164122.1 |
- |
2, 3 |
252 |
|
AK167970.1 |
- |
2, 3 |
278 |
|
AK168022.1 |
- |
2, 3 |
284 |
|
AK168678.1 |
- |
2, 3 |
355 |
|
AK168800.1 |
- |
2, 3 |
263 |
|
BC008622.1 |
- |
2, 3 |
285 |
3588758 |
BC010224.1 |
- |
2, 3 |
287 |
3500685 |
BY141813.1 |
- |
2, 3 |
313 |
|
View Full Product Details