Gene Transcripts
- Gene Symbol
- Smpx
- Entrez Gene ID
- 66106
- Gene Name
- small muscle protein, X-linked
- Gene Aliases
- 1010001C09Rik, Csl
- Chromosome Location
- Chr. X: 157698973 - 157752591 on Build GRCm38
- UniGene ID
- Mm.140340
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252591.2 |
NP_001239520.1
|
2, 3 |
408 |
|
GenBank mRNA |
AA434782.1 |
- |
2, 3 |
263 |
|
AF364070.1 |
- |
2, 3 |
247 |
|
AI035961.1 |
- |
2, 3 |
222 |
|
AJ245772.1 |
- |
2, 3 |
263 |
|
AK003105.1 |
- |
2, 3 |
255 |
|
AK009857.1 |
- |
2, 3 |
255 |
|
AK010172.1 |
- |
2, 3 |
255 |
|
AY026524.1 |
- |
2, 3 |
254 |
|
BC051197.1 |
- |
2, 3 |
207 |
1397202 |
BC061051.1 |
- |
2, 3 |
169 |
30300606 |
BC132358.1 |
- |
2, 3 |
83 |
40130635 |
BC138801.1 |
- |
2, 3 |
83 |
8861823 |
W18646.1 |
- |
3, 4 |
161 |
|
View Full Product Details