Gene Transcripts

Gene Symbol
Tma7
Entrez Gene ID
66167
Gene Name
translational machinery associated 7 homolog (S. cerevisiae)
Gene Aliases
1110017O22Rik, Ccdc72
Chromosome Location
Chr. 9: 109077988 - 109082381 on Build GRCm38
UniGene ID
Mm.2395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183250.2 NP_899073.1 3 161
GenBank mRNA AK002863.1 - 3 162
AK003762.1 - 3 162
AK160705.1 - 3 159
BC034602.1 - 3 143

View Full Product Details