Gene Transcripts

Gene Symbol
Med11
Entrez Gene ID
66172
Gene Name
mediator complex subunit 11
Gene Aliases
1110030J09Rik, AI465144, AW545069
Chromosome Location
Chr. 11: 70451931 - 70453727 on Build GRCm38
UniGene ID
Mm.46424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025397.2 NP_079673.2 2 249
GenBank mRNA AK003984.1 - 2 227
AK008153.1 - 2 267
BC025057.1 - 2 249 4238297

View Full Product Details