Gene Transcripts

Gene Symbol
Commd6
Entrez Gene ID
66200
Gene Name
COMM domain containing 6
Gene Aliases
1110059J08Rik, 1700063H17Rik
Chromosome Location
Chr. 14: 101633766 - 101640471 on Build GRCm38
UniGene ID
Mm.250474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033132.3 NP_001028304.1 3 234
NM_001168592.1 NP_001162064.1 2 222
GenBank mRNA AK004316.1 - 3 208
AK008220.1 - 3 216
AK009796.1 - 3 229
AK011226.1 - 3 219
AK018711.1 - 3 208
AK019351.1 - 3 210
AK019432.1 - 3 211
AK020234.1 - 3 207
AK034223.1 - 1 3482
AK045407.1 - 2708
BC132261.1 - 3 221 40130538
BC132265.1 - 3 221 40130542
BC145360.1 - 3 221 9053918
BC145361.1 - 2 209 9053919
CK621880.1 - 2 185

View Full Product Details