Gene Transcripts

Gene Symbol
LTA4H
Entrez Gene ID
4048
Gene Name
leukotriene A4 hydrolase
Gene Aliases
Chromosome Location
Chr. 12: 96000753 - 96043520 on Build GRCh38
UniGene ID
Hs.524648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000895.2 NP_000886.1 6 739
NM_001256643.1 NP_001243572.1 6 670
NM_001256644.1 NP_001243573.1 6 670
NR_132659.1 6 678
XM_005268871.1 XP_005268928.1 6 729
XM_011538348.1 XP_011536650.1 6 729
XM_011538349.2 XP_011536651.1 6 729
GenBank mRNA AB209740.1 - 6 606
AK293630.1 - 6 658
AK298017.1 - 6 670
AK301308.1 - 6 548
AK308184.1 - 6 668
BC032528.1 - 6 656 5502553
BX647158.1 - 6 627
CR457068.1 - 6 597
DB019628.1 - 6 739
J02959.1 - 6 627
J03459.1 - 6 665

View Full Product Details