Gene Transcripts

Gene Symbol
Ergic3
Entrez Gene ID
66366
Gene Name
ERGIC and golgi 3
Gene Aliases
2310015B14Rik, AV318804, CGI-54, D2Ucla1, NY-BR-84, Sdbcag84
Chromosome Location
Chr. 2: 156008033 - 156018279 on Build GRCm38
UniGene ID
Mm.141276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025516.4 NP_079792.1 9, 10 839
GenBank mRNA AK009350.1 - 9, 10 840
AK013942.1 - 9, 10 839
AK050285.1 - 9, 10 854
BC043720.1 - 9, 10 864 5375947
BC057130.1 - 10, 11 891 6819420

View Full Product Details