Gene Transcripts

Gene Symbol
Commd5
Entrez Gene ID
66398
Gene Name
COMM domain containing 5
Gene Aliases
2310065H03Rik, AI854466, D15Ertd81e, Hcarg
Chromosome Location
Chr. 15: 76899941 - 76901297 on Build GRCm38
UniGene ID
Mm.291716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025536.2 NP_079812.1 2 656
GenBank mRNA AK010045.1 - 2 657
AK013148.1 - 1 1040
BC025891.1 - 2 621 5035059

View Full Product Details