Gene Transcripts

Gene Symbol
Ubr7
Entrez Gene ID
66622
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Gene Aliases
5730410I19Rik, AA589405, AW557761
Chromosome Location
Chr. 12: 102757975 - 102777701 on Build GRCm38
UniGene ID
Mm.34261
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025666.2 NP_079942.1 9 1150
GenBank mRNA AK088208.1 - 9 1152
AK146308.1 - 9 1152
AK149989.1 - 9 1105
AK155611.1 - 9 1021
BC051678.1 - 6 678
BC058535.1 - 8 834
BC079531.1 - 8 847
BC094323.1 - 9 1157
BC094423.1 - 9 1108 6812389

View Full Product Details