Gene Transcripts

Gene Symbol
Tanc1
Entrez Gene ID
66860
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases
1200003E16Rik, Tanc, mKIAA1728
Chromosome Location
Chr. 2: 59611848 - 59846213 on Build GRCm38
UniGene ID
Mm.27917
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290659.1 NP_001277588.1 9 1346
NM_198294.3 NP_938036.2 10 1522
GenBank mRNA AK030022.1 - 7 1126
BC079914.1 - 9 1325 30668628
BC141367.1 - 9 1336 9055980

View Full Product Details