Gene Transcripts
- Gene Symbol
- Peli1
- Entrez Gene ID
- 67245
- Gene Name
- pellino 1
- Gene Aliases
- 2810468L03Rik, A930031K15Rik, AA409794, AI586297, D11Ertd676e
- Chromosome Location
- Chr. 11: 21091324 - 21150327 on Build GRCm38
- UniGene ID
- Mm.28957
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023324.2 |
NP_075813.2
|
2, 3 |
490 |
|
GenBank mRNA |
AF302503.1 |
- |
1, 2 |
712 |
|
AK020915.1 |
- |
2, 3 |
524 |
|
AK029586.1 |
- |
2, 3 |
388 |
|
AK151660.1 |
- |
2, 3 |
491 |
|
AK152900.1 |
- |
2, 3 |
491 |
|
AK152997.1 |
- |
2, 3 |
491 |
|
AK153372.1 |
- |
2, 3 |
491 |
|
AK167525.1 |
- |
2, 3 |
491 |
|
BC016515.1 |
- |
1, 2 |
89 |
3600130 |
BC026695.1 |
- |
2, 3 |
454 |
|
View Full Product Details