Gene Transcripts
- Gene Symbol
- Rpl37
- Entrez Gene ID
- 67281
- Gene Name
- ribosomal protein L37
- Gene Aliases
- 3110005M08Rik
- Chromosome Location
- Chr. 15: 5116613 - 5119140 on Build GRCm38
- UniGene ID
- Mm.10474
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026069.3 |
NP_080345.1
|
3, 4 |
290 |
|
GenBank mRNA |
AK002592.1 |
- |
3, 4 |
259 |
|
AK008563.1 |
- |
3, 4 |
259 |
|
AK011101.1 |
- |
3, 4 |
251 |
|
AK012544.1 |
- |
3, 4 |
259 |
|
AK014003.1 |
- |
3, 4 |
259 |
|
AK019020.1 |
- |
3, 4 |
259 |
|
AK019294.1 |
- |
3, 4 |
257 |
|
AK028138.1 |
- |
3, 4 |
249 |
|
AK168550.1 |
- |
3, 4 |
259 |
|
BC034524.1 |
- |
|
237 |
|
BC054388.1 |
- |
3, 4 |
246 |
6478626 |
BC081438.1 |
- |
2, 3 |
221 |
6817584 |
- Gene Symbol
- Rpl37rt
- Entrez Gene ID
- 100502825
- Gene Name
- ribosomal protein L37, retrotransposed
- Gene Aliases
- Gm13826, OTTMUSG00000014624
- Chromosome Location
- Chr. 5: 115102922 - 115110268 on Build GRCm38
- UniGene ID
- Mm.361657
- Species
- Mus musculus
View Full Product Details