Gene Transcripts
- Gene Symbol
- Parp6
- Entrez Gene ID
- 67287
- Gene Name
- poly (ADP-ribose) polymerase family, member 6
- Gene Aliases
- 1700119G14Rik, 2310028P13Rik, 3110038K10Rik, ARTD17, C030013N01Rik, PARP-6
- Chromosome Location
- Chr. 9: 59617284 - 59650290 on Build GRCm38
- UniGene ID
- Mm.439943
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205239.1 |
NP_001192168.1
|
15, 16 |
1758 |
|
NM_029922.3 |
NP_084198.2
|
15, 16 |
1698 |
|
GenBank mRNA |
AK009530.1 |
- |
5, 6 |
458 |
|
AK014142.1 |
- |
6, 7 |
501 |
|
AK035349.1 |
- |
15, 16 |
1622 |
|
AK075876.1 |
- |
5, 6 |
460 |
|
AK076321.1 |
- |
15, 16 |
1622 |
|
BC030184.1 |
- |
1, 2 |
48 |
|
BC031387.1 |
- |
14, 15 |
1429 |
5038718 |
BC052064.1 |
- |
15, 16 |
1696 |
|
BC058266.1 |
- |
6, 7 |
652 |
|
BC062096.1 |
- |
14, 15 |
1615 |
5716648 |
View Full Product Details