Gene Transcripts

Gene Symbol
Hoga1
Entrez Gene ID
67432
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
0610010D20Rik, Dhdpsl, Npl2
Chromosome Location
Chr. 19: 42045851 - 42070939 on Build GRCm38
UniGene ID
Mm.24196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026152.1 NP_080428.1 1 134
GenBank mRNA AK002457.1 - 1 134
BC016430.1 - 1 103 4235003

View Full Product Details