Gene Transcripts

Gene Symbol
MAP6
Entrez Gene ID
4135
Gene Name
microtubule associated protein 6
Gene Aliases
MAP6-N, MTAP6, N-STOP, STOP
Chromosome Location
Chr. 11: 75586914 - 75671108 on Build GRCh38
UniGene ID
Hs.585540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033063.1 NP_149052.1 2 1135
NM_207577.1 NP_997460.1 2 1135
XM_006718556.3 XP_006718619.1 2 3774
XM_011545039.2 XP_011543341.1 2 3775
XM_017017755.1 XP_016873244.1 2 3674
XM_017017756.1 XP_016873245.1 2 237
XM_017017757.1 XP_016873246.1 3 304
XM_017017758.1 XP_016873247.1 2 271
GenBank mRNA AB058781.1 - 2 1135
AK123340.1 - 2 243
BC063860.1 - 2 801
BC139780.1 - 3 374 40122938
BC150254.1 - 2 1135 9007181

View Full Product Details