Gene Transcripts

Gene Symbol
Larp6
Entrez Gene ID
67557
Gene Name
La ribonucleoprotein domain family, member 6
Gene Aliases
5430431G03Rik, AI552438, Achn
Chromosome Location
Chr. 9: 60713121 - 60738801 on Build GRCm38
UniGene ID
Mm.44546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026235.4 NP_080511.2 3 1036
GenBank mRNA AK017372.1 - 3 1041
AK035809.1 - 613
AK039614.1 - 3 1019
AK047452.1 - 1 340
AK087521.1 - 3 1039
BC090615.1 - 3 1168 6809572

View Full Product Details