Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026261.2 | NP_080537.1 | 1 | 600 | |
GenBank mRNA | AK006408.1 | - | 1 | 526 | |
AK015868.1 | - | 1 | 601 | ||
AK016746.1 | - | 1 | 557 | ||
BC048443.1 | - | 1 | 539 | 6775231 | |
BC061071.1 | - | 1 | 565 | 6774419 |