Gene Transcripts

Gene Symbol
Aasdhppt
Entrez Gene ID
67618
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5
Chromosome Location
Chr. 9: 4294793 - 4309509 on Build GRCm38
UniGene ID
Mm.33970
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026276.3 NP_080552.3 2 359
GenBank mRNA AK007737.1 - 2 360
AK008554.1 - 2 324
AK010557.1 - 2 319
AK013111.1 - 2 313
AK014188.1 - 2 302
AK028700.1 - 2 316
AK029523.1 - 2 301
AK031766.1 - 2 303
AK076099.1 - 2 314
AK076866.1 - 2 333
AK081818.1 - 2 335
AK088977.1 - 2 299
AK144671.1 - 2 333
AK214035.1 - 1 27
AK215948.1 - 1 27
BC030043.1 - 2 294 1348651
BC038013.1 - 2 330 3497754
BC049851.1 - 2 320 3481972

View Full Product Details