Gene Transcripts

Gene Symbol
Enoph1
Entrez Gene ID
67870
Gene Name
enolase-phosphatase 1
Gene Aliases
2310057D15Rik, BB183658, C81437
Chromosome Location
Chr. 5: 100039994 - 100068925 on Build GRCm38
UniGene ID
Mm.11311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163035.1 NP_001156507.1 5 805
NM_026421.3 NP_080697.2 5 805
GenBank mRNA AK009954.1 - 5 788
AK079062.1 - 5 799
AK080714.1 - 5 756
AK082747.1 - 5 780
AK152021.1 - 5 780
AK159356.1 - 5 778
AK167575.1 - 5 816
AK168696.1 - 5 674
BC021429.1 - 5 732 5030617

View Full Product Details