Gene Transcripts

Gene Symbol
Hcfc2
Entrez Gene ID
67933
Gene Name
host cell factor C2
Gene Aliases
1700129L13Rik, AU019838
Chromosome Location
Chr. 10: 82697497 - 82795865 on Build GRCm38
UniGene ID
Mm.127405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081218.1 NP_001074687.1 12, 13 1800
GenBank mRNA AK007317.1 - 3, 4 283
BC116411.1 - 5, 6 592
BC127176.1 - 5, 6 592

View Full Product Details