Gene Transcripts

Gene Symbol
Hormad1
Entrez Gene ID
67981
Gene Name
HORMA domain containing 1
Gene Aliases
4921522K05Rik, Nohma
Chromosome Location
Chr. 3: 95559640 - 95587940 on Build GRCm38
UniGene ID
Mm.179050
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289532.1 NP_001276461.1 6, 7 368
NM_001289534.1 NP_001276463.1 6, 7 368
NM_001289537.1 NP_001276466.1 6, 7 368
NM_026489.3 NP_080765.1 6, 7 359
GenBank mRNA AK014945.1 - 6, 7 367
AK016324.1 - 6, 7 367
AK016743.1 - 6, 7 362
AK163995.1 - 6, 7 367
AY626343.1 - 6, 7 367
AY634284.1 - 6, 7 367
BC051129.1 - 6, 7 367 6706580

View Full Product Details