Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026500.3 | NP_080776.1 | 2 | 710 | |
GenBank mRNA | AK004833.1 | - | 2 | 711 | |
AK013179.1 | - | 2 | 672 | ||
AK076462.1 | - | 2 | 665 | ||
AK133731.1 | - | 2 | 672 | ||
BC023840.1 | - | 2 | 637 | 5344890 |