Gene Transcripts

Gene Symbol
Mpnd
Entrez Gene ID
68047
Gene Name
MPN domain containing
Gene Aliases
E130307M08Rik
Chromosome Location
Chr. 17: 56009157 - 56016790 on Build GRCm38
UniGene ID
Mm.358710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026530.5 NP_080806.4 11 1319
GenBank mRNA AK012128.1 - 10 1274
AK146973.1 - 11 1304
AK160356.1 - 11 1288
AK172381.1 - 10 1062
AK212460.1 - 1 44
BC017625.1 - 10 1268

View Full Product Details