Gene Transcripts
- Gene Symbol
- Aen
- Entrez Gene ID
- 68048
- Gene Name
- apoptosis enhancing nuclease
- Gene Aliases
- 2700083B06Rik, Isg20l1
- Chromosome Location
- Chr. 7: 78895927 - 78908833 on Build GRCm38
- UniGene ID
- Mm.34109
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001162939.1 |
NP_001156411.1
|
2, 3 |
572 |
|
NM_026531.4 |
NP_080807.3
|
2, 3 |
686 |
|
GenBank mRNA |
AK012555.1 |
- |
2, 3 |
671 |
|
AK031972.1 |
- |
2, 3 |
687 |
|
AK089090.1 |
- |
2, 3 |
508 |
|
AK090056.1 |
- |
2, 3 |
555 |
|
AK165708.1 |
- |
2, 3 |
566 |
|
AK166806.1 |
- |
2, 3 |
549 |
|
AK167042.1 |
- |
2, 3 |
549 |
|
AK167658.1 |
- |
2, 3 |
645 |
|
BC022614.1 |
- |
2, 3 |
766 |
4167800 |
View Full Product Details