Gene Transcripts

Gene Symbol
Aph1c
Entrez Gene ID
68318
Gene Name
anterior pharynx defective 1c homolog (C. elegans)
Gene Aliases
0610008A10Rik, APH-1b, Aph1b
Chromosome Location
Chr. 9: 66814994 - 66834779 on Build GRCm38
UniGene ID
Mm.284229
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026674.3 NP_080950.1 2 287
GenBank mRNA AB197123.1 - 2 107
AK002310.1 - 2 253
AK080464.1 - 1 234
AK155147.1 - 2 301
BC050923.1 - 2 287 6410104
BC063254.1 - 2 287 6410104

View Full Product Details