Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164742.1 | NP_001158214.1 | 3 | 743 | |
NM_026687.2 | NP_080963.2 | 4 | 1048 | ||
GenBank mRNA | AB233454.1 | - | 4 | 1048 | |
AK002574.1 | - | 3 | 936 | ||
AK165409.1 | - | 3 | 721 | ||
BC119526.1 | - | 3 | 876 | 40056096 |