Gene Transcripts

Gene Symbol
Ankrd13d
Entrez Gene ID
68423
Gene Name
ankyrin repeat domain 13 family, member D
Gene Aliases
0710001P18Rik, AI430801
Chromosome Location
Chr. 19: 4270180 - 4283482 on Build GRCm38
UniGene ID
Mm.23642
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026720.2 NP_080996.2 8 1036
GenBank mRNA AK089186.1 - 8 1029
BC058982.1 - 8 947 5685059

View Full Product Details