Gene Transcripts

Gene Symbol
Krtap6-5
Entrez Gene ID
68484
Gene Name
keratin associated protein 6-5
Gene Aliases
1110002D05Rik, Krtap16-8
Chromosome Location
Chr. 16: 89047289 - 89047899 on Build GRCm38
UniGene ID
Mm.46209
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130856.2 NP_570926.1 1 535
GenBank mRNA AF345298.1 - 1 517
AK003281.1 - 1 536
BC059086.1 - 1 519

View Full Product Details