Gene Transcripts

Gene Symbol
Tspan11
Entrez Gene ID
68498
Gene Name
tetraspanin 11
Gene Aliases
1110014F12Rik, AU067806
Chromosome Location
Chr. 6: 127887622 - 127953031 on Build GRCm38
UniGene ID
Mm.174368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026743.3 NP_081019.1 6 776
GenBank mRNA AK003691.1 - 6 628
AK028808.1 - 6 667
AK053788.1 - 6 763
AK087294.1 - 6 792
AK090022.1 - 6 776
BC057639.1 - 6 618 4218418

View Full Product Details