Gene Transcripts

Gene Symbol
Ppfia4
Entrez Gene ID
68507
Gene Name
protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 4
Gene Aliases
1110008G13Rik, AI448359, AI852265, C81506
Chromosome Location
Chr. 1: 134296783 - 134344750 on Build GRCm38
UniGene ID
Mm.295105
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144855.1 NP_001138327.1 18 2791
GenBank mRNA AK173053.1 - 18 2281
EU568872.1 - 18 2791

View Full Product Details