Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145920.3 | NP_666032.1 | 1, 2 | 261 | |
GenBank mRNA | AB083066.1 | - | 1, 2 | 191 | |
AK044977.1 | - | 1, 2 | 132 | ||
AK134298.1 | - | 1, 2 | 244 | ||
AK165051.1 | - | 1, 2 | 146 | ||
BC037473.1 | - | 1, 2 | 242 | 4237472 | |
BC064473.1 | - | 1, 2 | 252 | 3156380 | |
BY270131.1 | - | 1, 2 | 262 |