Gene Transcripts

Gene Symbol
Gtf2f2
Entrez Gene ID
68705
Gene Name
general transcription factor IIF, polypeptide 2
Gene Aliases
1110031C13Rik, AI326315
Chromosome Location
Chr. 14: 75896937 - 76010865 on Build GRCm38
UniGene ID
Mm.20415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026816.3 NP_081092.1 3, 4 277
GenBank mRNA AK144858.1 - 3, 4 278
BC027173.1 - 3, 4 273 3986466

View Full Product Details