Gene Transcripts

Gene Symbol
Foxk2
Entrez Gene ID
68837
Gene Name
forkhead box K2
Gene Aliases
1110054H05Rik, 5730434B08Rik, 6230415M23Rik, ILF, Ilf1
Chromosome Location
Chr. 11: 121259987 - 121309896 on Build GRCm38
UniGene ID
Mm.209750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080932.2 NP_001074401.2 7, 8 1699
GenBank mRNA EU882160.1 - 7, 8 1699

View Full Product Details