Gene Transcripts

Gene Symbol
Ubap2
Entrez Gene ID
68926
Gene Name
ubiquitin-associated protein 2
Gene Aliases
1190005K07Rik, AA408600, AU045235, UBAP-2, mKIAA1491
Chromosome Location
Chr. 4: 41194313 - 41275321 on Build GRCm38
UniGene ID
Mm.41864
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026872.1 NP_081148.1 15 1839
GenBank mRNA AF276965.1 - 14 1743
BC005482.1 - 2 410
BC007179.1 - 15 1839 3496608

View Full Product Details