Gene Transcripts

Gene Symbol
Pink1
Entrez Gene ID
68943
Gene Name
PTEN induced putative kinase 1
Gene Aliases
1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387
Chromosome Location
Chr. 4: 138313409 - 138326296 on Build GRCm38
UniGene ID
Mm.18539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026880.2 NP_081156.2 4 1020
GenBank mRNA AB053476.1 - 4 952
AK050322.1 - 4 1000
AK155474.1 - 4 1006
AK165480.1 - 4 999
AK180089.1 - 1 81
AK198589.1 - 1 82
BC018594.1 - 4 983
BC067066.1 - 4 988 30542123

View Full Product Details