Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163421.1 | NP_001156893.1 | 6 | 454 | |
NM_001163422.1 | NP_001156894.1 | 6 | 454 | ||
NM_026895.1 | NP_081171.1 | 6 | 454 | ||
GenBank mRNA | AK005193.1 | - | 6 | 425 | |
AK156071.1 | - | 6 | 405 | ||
BC061248.1 | - | 6 | 426 |